MAS™ Urinalysis Controls
Actual product may vary
Thermo Scientific™
MAS™ Urinalysis Controls
Combine dipstick methods and microscopic components with bi-level, liquid assayed MAS Urinalysis Controls, assayed for most reagent strips and strip systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevelQuantity
024224315 mL
024222115 mL
024225160 mL
024227360 mL
Catalog number 024224
Price (USD)
-
Level:
3
Quantity:
15 mL
Combine dipstick methods and microscopic components with bi-level, liquid assayed Thermo Scientific™ MAS™ Urinalysis Controls. This control is assayed for most major reagent strips and strip systems.
  • Human urine matrix
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionUrinalysis Control, Assayed
Level3
Packaging4 Pk.
Quantity15 mL
Shelf Life12 Months
TypeControl
Detectable AnalytesBilirubin, Blood, Creatinine, Crystals, Glucose, hCG, Ketones, Leukocyte Esterase, Microalbumin, Nitrite, Osmolality, pH, Potassium, Protein, Red Blood Cells, Sodium, Specific Gravity, Urobilinogen, White Blood Cells
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C