Orthophosphoric acid, ≥85.0-88.0%, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Orthophosphoric acid, ≥85.0-88.0%, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets EP, BP, and NF specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
4 L
700 lb.
Packaging:
View all
Poly bottle
Catalog number 0268-06
Price (USD)
-
Quantity:
4 L
Packaging:
Poly bottle
This product is GMP Manufactured and meets EP, BP, and NF specifications.
This product is GMP Manufactured and meets EP, BP, and NF specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS7664-38-2
Chemical Name or MaterialOrthophosphoric acid
ColorColorless
Melting Point28°C
Molecular FormulaH3PO4
PackagingPoly bottle
SynonymPhosphoric acid
GradeMulti-Compendial
Molecular Weight (g/mol)98
Percent Purity85 to 88%
Quantity4 L
Unit SizeEach