Al-23 Tube, One End Closed - FAQs
View additional product information for Al-23 Tube, One End Closed - FAQs (030069.QAT)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.