Di-tert-butyl phosphite, 95%
Di-tert-butyl phosphite, 95%
Di-tert-butyl phosphite, 95%
Thermo Scientific Chemicals

Di-tert-butyl phosphite, 95%

As a solvent, as an antioxidant, and as an intermediate | CAS: 13086-84-5 | C8H18O3P | 193.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 030233.03
also known as 030233-03
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.80
Save 6.15 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.80
Save 6.15 (15%)
Each
Chemical Identifiers
CAS13086-84-5
IUPAC Namebis(tert-butoxy)(oxo)-λ⁵-phosphanylium
Molecular FormulaC8H18O3P
InChI KeyGEBLOQXLELCEEO-UHFFFAOYSA-N
SMILESCC(C)(C)O[P+](=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (unspecified)> 94.0%
Di-tert-butyl phosphite is used as a solvent, as an antioxidant, and as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only