Tricyclohexylphosphine oxide
Tricyclohexylphosphine oxide
Tricyclohexylphosphine oxide
Thermo Scientific Chemicals

Tricyclohexylphosphine oxide

CAS: 13689-19-5 | C18H33OP | 296.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 030387.14
also known as 030387-14
Price (USD)
488.65
Online Exclusive
543.00
Save 54.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
488.65
Online Exclusive
543.00
Save 54.35 (10%)
Each
Specifications
CAS13689-19-5
Chemical Name or MaterialTricyclohexylphosphine oxide
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
Formula Weight296.42
Health Hazard 1H301-H315-H319-H335
View more
Tricyclohexylphosphine oxide is used as a ligand for a range of metal-catalyzed organic transformations. Ligand used in the Pd-catalyzed coupling of malononitrile with aryl halides.1 Bulky phosphine ligand used with a Pd(0)-triolefinic macrocycle catalyst for Suzuki coupling of aryl bromides and chlorides. This ligand is applied with Ni as a key intermediate in the formation of cylcopentane compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only