Triethylphosphine oxide
Triethylphosphine oxide
Triethylphosphine oxide
Thermo Scientific Chemicals

Triethylphosphine oxide

CAS: 597-50-2 | C6H15OP | 134.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 030391.06
also known as 030391-06
Price (USD)
288.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
288.00
Each
Chemical Identifiers
CAS597-50-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals
Assay (GC)≥97.5%
Triethylphosphine oxide is used as a probe molecule and systems are evaluated by 31P NMR spectroscopy. Triethylphosphine oxide is used to produce tris(perfluoroethyl)difluorophosphane. This reaction will need reagent of HF. This reaction needs electrochemical fluorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only