Trimethylphosphine oxide
Trimethylphosphine oxide
Trimethylphosphine oxide
Thermo Scientific Chemicals

Trimethylphosphine oxide

CAS: 676-96-0 | C3H9OP | 92.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 030394.03
also known as 030394-03
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS676-96-0
IUPAC Name(dimethylphosphoryl)methane
Molecular FormulaC3H9OP
InChI KeyLRMLWYXJORUTBG-UHFFFAOYSA-N
SMILESCP(C)(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)White
Assay from Supplier's CofA≥95.0%
Trimethylphosphine oxide is a common side product in organic synthesis, including the Wittig, Staudinger, Mitsunobu reactions and also in the manufacture of triphenylphosphine. It is a popular reagent to induce the crystallization of various chemical compounds. It acts as an excellent ligand for hard metal centers. It can react with 9-methyl-9-azonianthracene methyl sulfate to get (10-methyl-9,10-dihydro-acridin-9-yl)-phosphonic acid dimethyl ester using sodium iodide and acetonitrile as solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only