Calcium nitrate tetrahydrate, 99.98% (metals basis)
Calcium nitrate tetrahydrate, 99.98% (metals basis)
Calcium nitrate tetrahydrate, 99.98% (metals basis)
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 99.98% (metals basis)

CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 030482.14
also known as 030482-14
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.70
Save 15.05 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.70
Save 15.05 (15%)
Each
Chemical Identifiers
CAS13477-34-4
Molecular FormulaCaH8N2O10
InChI KeyMWGCGFYACDTFSB-UHFFFAOYSA-N
SMILESO.[Ca++].[O-][N+]([O-])=O
Molecular Weight (g/mol)236.15
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.02% max.
Calcium nitrate tetrahydrate is used as a nitrogenous fertilizer and a corrosive inhibitor in diesel fuels. It is utilized in plant cells for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only