Lithium 2,4-pentanedionate, 99.5%
Lithium 2,4-pentanedionate, 99.5%
Lithium 2,4-pentanedionate, 99.5%
Thermo Scientific Chemicals

Lithium 2,4-pentanedionate, 99.5%

CAS: 18115-70-3 | C5H7LiO2 | 106.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 030529.09
also known as 030529-09
Price (USD)
25.80
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
25.80
Each
Chemical Identifiers
CAS18115-70-3
IUPAC Namelithium(1+) (2Z)-4-oxopent-2-en-2-olate
Molecular FormulaC5H7LiO2
InChI KeyJTEOOCAGEXVCBQ-LNKPDPKZSA-M
SMILES[Li+].C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder or crystals or granules
Appearance (Color)White to pale yellow
Assay from Suppliers CofA≥97.5%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only