Tin(IV) chloride bis(2,4-pentanedionate), 95%
Tin(IV) chloride bis(2,4-pentanedionate), 95%
Tin(IV) chloride bis(2,4-pentanedionate), 95%
Thermo Scientific Chemicals

Tin(IV) chloride bis(2,4-pentanedionate), 95%

CAS: 16919-46-3 | C10H14Cl2O4Sn | 387.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 030532.09
also known as 030532-09
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
142.65
Online Exclusive
158.00
Save 15.35 (10%)
Each
Specifications
CAS16919-46-3
Chemical Name or MaterialTin(IV) chloride bis(2,4-pentanedionate)
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECS Number240-972-5
Formula Weight387.83
View more
It is used as a raw material in synthesis. It is also used as NMR shift reagents and as catalysts for organic synthesis, and precursors to industrial hydroformylation catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only