Thermo Scientific Chemicals

Strontium nitrate, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: 030534.D9
5 x 1 kg, Each
Thermo Scientific Chemicals

Strontium nitrate, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: 030534.D9
5 x 1 kg, Each
Quantity
Catalog number: 030534.D9
also known as 030534-D9
Price (USD)
Price: 261.00
Online price: 221.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10042-76-9
IUPAC Name
strontium(2+) dinitrate
Molecular Formula
N2O6Sr
InChI Key
DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES
[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Assay (unspecified)
97.6% min

Description

In railroad flares, signal lights, marine signalsStrontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. Mixed with glycolic acid, it is used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text