2,2'-Bipyridine, ACS, 98%
2,2'-Bipyridine, ACS, 98%
2,2'-Bipyridine, ACS, 98%
Thermo Scientific Chemicals

2,2'-Bipyridine, ACS, 98%

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 030569.18
also known as 030569-18
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS366-18-7
IUPAC Name2,2'-bipyridine
Molecular FormulaC10H8N2
InChI KeyROFVEXUMMXZLPA-UHFFFAOYSA-N
SMILESC1=CC=C(N=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentClarity and absorbance of solution P.T.
CommentSensitivity to ferrous iron P.T.
Melting Point≥70?
2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only