Lithium tri-tert-butoxyaluminum hydride, 94%
Lithium tri-tert-butoxyaluminum hydride, 94%
Lithium tri-tert-butoxyaluminum hydride, 94%
Thermo Scientific Chemicals

Lithium tri-tert-butoxyaluminum hydride, 94%

CAS: 17476-04-9
Have Questions?
Catalog number 030716.09
also known as 030716-09
Price (USD)
-
Request bulk or custom format
Chemical Identifiers
CAS17476-04-9
IUPAC Namealuminium(3+) tris(2-methylpropan-2-olate) lithium
Molecular FormulaC12H27AlLiO3
InChI KeyHTBVGZAVHBZXMS-UHFFFAOYSA-N
SMILES[Li].[Al+3].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or chunks
Gas Volumetric assayHydrogen evolution: 92.5-107.5%
Infrared spectrumConforms
Lithium aluminum-tri-tert-butoxyhydride is used as a reducing agent. It is very useful in selective reductions of acyl halides, aldehydes, ketones, and esters, it is also used as an antimicrobial, cholesterol lowering agent, for ion-permeable synthetic membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only