Salicylic acid, ACS, 99+%
Salicylic acid, ACS, 99+%
Salicylic acid, ACS, 99+%
Thermo Scientific Chemicals

Salicylic acid, ACS, 99+%

CAS: 69-72-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number 030782.36
also known as 030782-36
Price (USD)
80.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
80.00
Each
Chemical Identifiers
CAS69-72-7
IUPAC Name2-hydroxybenzoic acid
Molecular FormulaC7H6O3
InChI KeyYGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.01%
Impurity contentCl 0.001%
Assay (HPLC)> 99.0% C7H6O3
Impurity contentFe 2ppm
Impurity contentSO₄ 0.003%
View more
Salicylic acid is used as a food preservative as well as used in the treatment of many skin disorders such as acne, dandruff, psoriasis and seborrheic dermatitis of the skin and scalp. It is an antiseptic. It is mainly used in the pharmaceutical industry for the manufacture of antipyretic, analgesic, anti-inflammatory and diuretic drugs. It is also used in the dye industry for making azo direct dyes and acidic mordant dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only