Tetra-n-butylammonium perchlorate, 99+%
Tetra-n-butylammonium perchlorate, 99+%
Tetra-n-butylammonium perchlorate, 99+%
Thermo Scientific Chemicals

Tetra-n-butylammonium perchlorate, 99+%

CAS: 1923-70-2 | C16H36ClNO4 | 341.917 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
25 g
100 g
Catalog number 030801.14
also known as 030801-14
Price (USD)
113.65
Online exclusive
126.00
Save 12.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
113.65
Online exclusive
126.00
Save 12.35
Each
Chemical Identifiers
CAS1923-70-2
IUPAC Nametetrabutylazanium perchlorate
Molecular FormulaC16H36ClNO4
InChI KeyKBLZDCFTQSIIOH-UHFFFAOYSA-M
SMILES[O-][Cl](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formcrystalline powder
Assay (Titration)>/= 99.0%
Tetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only