2-Butanone, 97%
2-Butanone, 97%
2-Butanone, 97%
Thermo Scientific Chemicals

2-Butanone, 97%

CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 031069.M1
also known as 031069-M1
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Add to cart
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS78-93-3
IUPAC Namebutan-2-one
Molecular FormulaC4H8O
InChI KeyZWEHNKRNPOVVGH-UHFFFAOYSA-N
SMILESCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale brown
FormLiquid
Assay (GC)≥97%
2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only