Iron(III) phosphate hydrate
Iron(III) phosphate hydrate
Iron(III) phosphate hydrate
Iron(III) phosphate hydrate
Thermo Scientific Chemicals

Iron(III) phosphate hydrate

CAS: 51833-68-2 | FeH2O5P | 168.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 031106.A1
also known as 031106-A1
Price (USD)
190.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
190.00
Each
Chemical Identifiers
CAS51833-68-2
IUPAC Nameiron(3+) hydrate phosphate
Molecular FormulaFeH2O5P
InChI KeyRQKNQWUJROKKEQ-UHFFFAOYSA-K
SMILESO.[Fe+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)26.0-32.0% Fe

Poly(methyl methacrylate) is used in the lenses of exterior lights of automobile and automotive components such as badges, steering wheel, tachometer covers, warning triangles and fascia panels. It is in the form of sheets used in shatter resistant panels for building windows, skylights, bullet proof security barriers, bathtubs, LCD screens, furniture and many other applications. It is used as the matrix in laser dye-doped organic solid-state gain media for tunable solid state dye lasers. Further, it is used as a shield to stop beta radiation emitted from radioisotopes, as dosimeter devices during the gamma irradiation process and as a dispersant for ceramic powders to stabilize colloidal suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only