Potassium trioxalatoferrate(III) trihydrate
Potassium trioxalatoferrate(III) trihydrate
Potassium trioxalatoferrate(III) trihydrate
Thermo Scientific Chemicals

Potassium trioxalatoferrate(III) trihydrate

CAS: 5936-11-8 | C6H3FeK3O12 | 440.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 031124.06
also known as 031124-06
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.80
Save 6.15 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.80
Save 6.15 (14%)
Each
Specifications
CAS5936-11-8
Chemical Name or MaterialPotassium trioxalatoferrate(III) trihydrate
ColorGreen
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Density2.133 g/mL
View more
Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only