Iron(II) chloride, anhydrous, 99.5% (metals basis)
Iron(II) chloride, anhydrous, 99.5% (metals basis)
Iron(II) chloride, anhydrous, 99.5% (metals basis)
Thermo Scientific Chemicals

Iron(II) chloride, anhydrous, 99.5% (metals basis)

CAS: 7758-94-3 | Cl2Fe | 126.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Catalog number 031141.18
also known as 031141-18
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS7758-94-3
IUPAC Nameλ²-iron(2+) dichloride
Molecular FormulaCl2Fe
InChI KeyNMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
FormPowder
Total Metal Impurities≤0.5% (≤5000 ppm)
Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only