Iron(II) bromide, anhydrous, 98% min
Iron(II) bromide, anhydrous, 98% min
Iron(II) bromide, anhydrous, 98% min
Thermo Scientific Chemicals

Iron(II) bromide, anhydrous, 98% min

CAS: 7789-46-0 | Br2Fe | 215.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 031154.14
also known as 031154-14
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
230.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS7789-46-0
IUPAC Nameλ²-iron(2+) dibromide
Molecular FormulaBr2Fe
InChI KeyGYCHYNMREWYSKH-UHFFFAOYSA-L
SMILES[Fe++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Iron(II) bromide is used as an intermediate in the synthesis porphryins. It is also used as weak reducing agents. It is used as precursor for the preparation of [(C2H5)4N]2FeBr4 by reacting with tetraethylammonium bromide. It is also involved in the synthesis of iron pyrite nanocrystals and nanocrystalline thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only