1-Decene, 96%, remainder isomers
1-Decene, 96%, remainder isomers
1-Decene, 96%, remainder isomers
Thermo Scientific Chemicals

1-Decene, 96%, remainder isomers

CAS: 872-05-9 | C10H20 | 140.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 031213.A3
also known as 031213-A3
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
146.00
Save 21.35 (15%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
124.65
Special offer
Online exclusive
Ends: 30-Jun-2026
146.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS872-05-9
IUPAC Namedec-1-ene
Molecular FormulaC10H20
InChI KeyAFFLGGQVNFXPEV-UHFFFAOYSA-N
SMILESCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥95.0%
FormLiquid
1-Decene is the only isomer of industrial importance. it is used as a monomer in copolymers and is an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics. 1-Decene has been isolated from the leaves and rhizome of the plant Farfugium japonicum and has been detected as the initial product in the microbial degradation of n-decane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only