Dodecylbenzene, mixture of isomers
Dodecylbenzene, mixture of isomers
Dodecylbenzene, mixture of isomers
Dodecylbenzene, mixture of isomers
Thermo Scientific Chemicals

Dodecylbenzene, mixture of isomers

CAS: 123-01-3 | C18H30 | 246.438 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 031362.A1
also known as 031362-A1
Price (USD)
363.65
Online Exclusive
404.00
Save 40.35 (10%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
363.65
Online Exclusive
404.00
Save 40.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS123-01-3
IUPAC Namedodecylbenzene
Molecular FormulaC18H30
InChI KeyKWKXNDCHNDYVRT-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Carbon number distribution (C9%): Report
FormLiquid
Appearance (Color)Colorless to yellow
Molecular Weight238-250 (average relative molecular weight)
1-Phenyldodecane acts as a pseudo-stationary phase (PSP) marker in the preparation of microemulsion, 1-Phenyldodecane was degraded to phenylacetic acid by an oil-degrading bacterium. it is used to prepare an anionic detergent dodecylbenzene sodium sulfonate. It is a petrochemical-derived substitute for the tallow and coconut oil used in making soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only