Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1;ID (mm), 0.6 - FAQs
Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1;ID (mm), 0.6 - FAQs
View additional product information for Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1;ID (mm), 0.6 - FAQs (031479.DM, 031479.DW, 031479.DR)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.