Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 2.7;ID (mm), 1.7 - FAQs
Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 2.7;ID (mm), 1.7 - FAQs
View additional product information for Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 2.7mm, I.D.: 1.7mm, Thermo Scientific Chemicals - FAQs (031492.DR, 031492.QAT)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.