Search
View additional product information for Al-23 Tube (both ends open);OD (mm), 25;ID (mm), 20 - FAQs (031573.DR, 031573.DW, 031573.EH)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.