Al-23 Tube (both ends open);OD (mm), 30;ID (mm), 25
Thermo Scientific Chemicals
Al-23 Tube (both ends open);OD (mm), 30;ID (mm), 25
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 mm
600 mm
Catalog number 031574.EH
also known as 031574-EH
Price (USD)
334.65
Special offer
Online exclusive
Ends: 28-Dec-2026
393.00
Save 58.35
Each
Quantity:
600 mm
Request bulk or custom format
Price (USD)
334.65
Special offer
Online exclusive
Ends: 28-Dec-2026
393.00
Save 58.35
Each
Specifications
Chemical Name or MaterialAl-23 Tube
Name NoteBoth ends open
Recommended StorageAmbient temperatures
TSCAU
Quantity600 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Frequently asked questions (FAQs)
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.