Search
View additional product information for Al-23 Tube (both ends open);OD (mm), 35;ID (mm), 29 - FAQs (031575.DM, 031575.EO, 031575.DW)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.