Al-24 Tube (both ends open);OD (mm), 30;ID (mm), 24 - FAQs
Al-24 Tube (both ends open);OD (mm), 30;ID (mm), 24 - FAQs
View additional product information for Al-24 Tube (both ends open);OD (mm), 30;ID (mm), 24 - FAQs (031606.EH, 031606.EO)
1 product FAQs found
What is the maximum working temperature of Al-24 Tubes?
The maximum working temperature for the Al-24 Tube is 1950 degrees C.