Al-23 Tube, Both Ends Open (Thin Wall);OD (mm), 6;ID (mm), 5 - FAQs
Al-23 Tube, Both Ends Open (Thin Wall);OD (mm), 6;ID (mm), 5 - FAQs
View additional product information for Al-23 Tube, Both Ends Open (Thin Wall);OD (mm), 6;ID (mm), 5 - FAQs (031706.DM, 031706.DR, 031706.QAT)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.