What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.
Search
View additional product information for Al-23 Tube, Both Ends Open (Thin Wall);OD (mm), 10;ID (mm), 9 - FAQs (031709.DR, 031709.DW, 031709.DM)
1 product FAQs found