Thermo Scientific Chemicals

Malonic acid, Reagent Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 031715.22
100 g, Each
Thermo Scientific Chemicals

Malonic acid, Reagent Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 031715.22
100 g, Each
Quantity
Catalog number: 031715.22
also known as 031715-22
Price (USD)

Chemical Identifiers

CAS
141-82-2
IUPAC Name
propanedioic acid
Molecular Formula
C3H4O4
InChI Key
OFOBLEOULBTSOW-UHFFFAOYSA-N
SMILES
OC(=O)CC(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (unspecified)
≥99.5%
Chloride content
≤0.05%
Residue on ignition
≤0.1%

Description

Malonic acid is used to prepare barbiturates. Its derivative diethylmalonate is used in Knoevenagel condensation reactions as well as reacts with acetone to form Meldrum's acid. It is used to control acidity, preservative additive for foods and as an excipient in pharmaceutical formulation. It acts as a precursor to polyesters and as an active component in alkyd resins. Further, it is used as a building block in chemical synthesis. In addition to this, it is used to prepare starch-based resin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text