Methanol, ACS, absolute, low acetone, 99.8+%
Methanol, ACS, absolute, low acetone, 99.8+%
Methanol, ACS, absolute, low acetone, 99.8+%
Thermo Scientific Chemicals

Methanol, ACS, absolute, low acetone, 99.8+%

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
4 x 1 L
Catalog number 031721.K7
also known as 031721-K7
Price (USD)
88.65
Online Exclusive
98.80
Save 10.15 (10%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.80
Save 10.15 (10%)
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.1%
Residue on Evaporation≤0.001%
Solution Testin water P.T.
Appearance (Color)Colorless
Color (APHA)≤10
View more
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only