2-Methyl-2-butene, tech. 90%, remainder mainly 2-methyl-1-butene
2-Methyl-2-butene, tech. 90%, remainder mainly 2-methyl-1-butene
2-Methyl-2-butene, tech. 90%, remainder mainly 2-methyl-1-butene
Thermo Scientific Chemicals

2-Methyl-2-butene, tech. 90%, remainder mainly 2-methyl-1-butene

CAS: 513-35-9 | C5H10 | 70.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 031853.36
also known as 031853-36
Price (USD)
210.65
Special offer
Online exclusive
Ends: 30-Jun-2026
248.00
Save 37.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
210.65
Special offer
Online exclusive
Ends: 30-Jun-2026
248.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS513-35-9
IUPAC Name2-methylbut-2-ene
Molecular FormulaC5H10
InChI KeyBKOOMYPCSUNDGP-UHFFFAOYSA-N
SMILESCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥88.0%
2-Methyl-2-butene is used to prepare 3-Brom-2.3-dimethyl-1.1-dicyan-butan in the presence of 2,2'-azobis-(2,4-dimethyl-4-methoxyvaleronitrile) as a catalyst. It is used as a precursor for peroxyacetyl nitrate for the calibration purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only