4-Methyl-1,10-phenanthroline
4-Methyl-1,10-phenanthroline
4-Methyl-1,10-phenanthroline
Thermo Scientific Chemicals

4-Methyl-1,10-phenanthroline

CAS: 31301-28-7 | C13H10N2 | 194.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 031888.03
also known as 031888-03
Price (USD)
74.65
Online Exclusive
83.10
Save 8.45 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
83.10
Save 8.45 (10%)
Each
Specifications
CAS31301-28-7
Chemical Name or Material4-Methyl-1,10-phenanthroline
Formula Weight194.24
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
4-methyl-1,10-phenanthroline can be used to produce 1,10-Phenanthrolin-4-aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only