Phthalocyanine
Phthalocyanine
Phthalocyanine
Thermo Scientific Chemicals

Phthalocyanine

CAS: 574-93-6 | C32H18N8 | 514.552 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 032073.03
also known as 032073-03
Price (USD)
37.20
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.20
Each
Chemical Identifiers
CAS574-93-6
IUPAC Name2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1,3,5,7,9,11,13,15,17,19,21(38),22,24,26,28,30(37),31,33,35-nonadecaene
Molecular FormulaC32H18N8
InChI KeyIEQIEDJGQAUEQZ-UHFFFAOYSA-N
SMILESN1C2=NC3=C4C=CC=CC4=C(N3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple
Elemental Analysis> 98.0%
Formpowder
Pthalocyanine is a common macrocylic blue-green dye. It is also used as a suitable laser dye metal. Phthalocyanines have long been examined as catalysts for redox reactions. Areas of interest are the oxygen reduction reaction and the sweetening of gas streams by removal of hydrogen sulfide. Phthalocyanine compounds have been investigated as donor materials in molecular electronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only