Sodium 1-Hexanesulfonate Monohydrate, 99%
Sodium 1-Hexanesulfonate Monohydrate, 99%
Sodium 1-Hexanesulfonate Monohydrate, 99%
Thermo Scientific Chemicals

Sodium 1-Hexanesulfonate Monohydrate, 99%

CAS: 207300-91-2 | C6H15NaO4S | 206.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 032190.09
also known as 032190-09
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.90
Save 14.25
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.90
Save 14.25
Each
Chemical Identifiers
CAS207300-91-2
IUPAC Namesodium hexane-1-sulfonate hydrate
Molecular FormulaC6H15NaO4S
InChI KeyCLCGFJYKZGFGSQ-UHFFFAOYSA-M
SMILESO.[Na+].CCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at200 nm: ≤ 0.300 AU
Optical Absorbance at210 nm: ≤0.100AU
Loss on Drying7 to 9%
Appearance (Color)White
Formpowder
View more
Sodium 1-hexanesulfonate monohydrate is an ion pair reagent, which is used for the analysis of small organic compounds, pharmaceutical products and metabolites using high-performance liquid chromatography (HPLC). It is an efficient catalyst for the preparation of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only