Thermo Scientific Chemicals

Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads, Thermo Scientific Chemicals

Catalog number: 032256.30
250 g, Each
Thermo Scientific Chemicals

Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads, Thermo Scientific Chemicals

Catalog number: 032256.30
250 g, Each
Quantity
Catalog number: 032256.30
Price (USD)
Price: 53.80
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
70955-01-0
IUPAC Name
Molecular sieves
Synonym
2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL Number
MFCD00131613
Moisture holding capacity
20 wt% water capacity, min
Appearance (Color)
Blue-Tan
Form
beads

Description

Molecular sieves, 4 Å… is used to absorb molecules as large as butane. It is utilized for the separation of natural gas, liquefied petroleum gas (LPG) and alkenes. It finds application in the refining and purification of liquid and gases. It is also used for drying and purification of air, inert gases and solvents. It is actively involved to remove methanol, carbon dioxide and ammonia from fluid streams. It plays a vital role to remove heavy metal ions available in water. It is also used as a soap forming agent and as a detergent auxiliary. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text