Benzyldimethylhexadecylammonium chloride, 95%
Benzyldimethylhexadecylammonium chloride, 95%
Benzyldimethylhexadecylammonium chloride, 95%
Thermo Scientific Chemicals

Benzyldimethylhexadecylammonium chloride, 95%

CAS: 122-18-9 | C25H46ClN | 396.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 032377.30
also known as 032377-30
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS122-18-9
IUPAC Namebenzyl(hexadecyl)dimethylazanium chloride
Molecular FormulaC25H46ClN
InChI KeySXPWTBGAZSPLHA-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay from Supplier's CofA≥94%
Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only