Calcium oxalate monohydrate, 99%
Calcium oxalate monohydrate, 99%
Calcium oxalate monohydrate, 99%
Thermo Scientific Chemicals

Calcium oxalate monohydrate, 99%

CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 032430.22
also known as 032430-22
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.60
Save 8.95 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.60
Save 8.95 (15%)
Each
Chemical Identifiers
CAS5794-28-5
IUPAC Namecalcium hydrate oxalate
Molecular FormulaC2H2CaO5
InChI KeyLQHWSGSWNOHVHO-UHFFFAOYSA-L
SMILESO.[Ca++].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofA≥98.5%
Calcium oxalate monohydrate is used as a precursor in the production of oxalic acid, organic oxalates, ceramic glazes, and for rare-earth-metal separation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only