Methanol, ultrapure, Spectrophotometric Grade, 99.8+%
Methanol, ultrapure, Spectrophotometric Grade, 99.8+%
Methanol, ultrapure, Spectrophotometric Grade, 99.8+%
Thermo Scientific Chemicals

Methanol, ultrapure, Spectrophotometric Grade, 99.8+%

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 4 L
Catalog number 032435.K2
also known as 032435-K2
Price (USD)
53.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
53.00
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at225nm: ≥80% transmittance [UK]
Optical Absorbance at240nm: ≥95% transmittance [UK]
Optical Absorbance at260nm: ≤0.01 AU [US]
Optical Absorbance at265-400nm: ≥99% transmittance [UK]
Residue on Evaporation≤5ppm [US] / ≤0.0001% (≤1ppm) [UK]
View more
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only