Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%
Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%
Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%
Thermo Scientific Chemicals

Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%

CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
4 x 1 L
Catalog number 032436.K7
also known as 032436-K7
Price (USD)
405.65
Online Exclusive
450.00
Save 44.35 (10%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
405.65
Online Exclusive
450.00
Save 44.35 (10%)
Each
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.07%
Residue on Evaporation4ppm
Pyridine is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only