Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene
Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene
Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene
Thermo Scientific Chemicals

Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 032440.K2
also known as 032440-K2
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.02%
Impurity contentFree halogens P.T.
Residue on Evaporation3ppm
Dichloromethane is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only