n-Pentane, Spectrophotometric Grade, 99+% min.
n-Pentane, Spectrophotometric Grade, 99+% min.
n-Pentane, Spectrophotometric Grade, 99+% min.
Thermo Scientific Chemicals

n-Pentane, Spectrophotometric Grade, 99+% min.

CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Catalog number 032449.M1
also known as 032449-M1
Price (USD)
198.00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
198.00
Each
Chemical Identifiers
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at220nm: ≤0.15AU
Optical Absorbance at245nm: ≤0.01AU
Residue on Evaporation≤5ppm
Appearance (Color)Clear colorless
FormLiquid
View more
n-Pentane is used in spectrophotometry and environmental testing. It is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only