n-Hexane, Spectrophotometric Grade, 95+%
n-Hexane, Spectrophotometric Grade, 95+%
n-Hexane, Spectrophotometric Grade, 95+%
Thermo Scientific Chemicals

n-Hexane, Spectrophotometric Grade, 95+%

CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 032454.K7
also known as 032454-K7
Price (USD)
182.00
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
182.00
Each
Chemical Identifiers
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.02%
Residue on Evaporation5ppm
Assay (GC)>/= 95.0%
Optical Absorbance at@ 250 nm 0.01 AU, max @220 nm 0.10 AU, max @210 0.30 AU, max @195 nm 1.00 AU, max
Appearance (Color)Colorless
View more
n-Hexane used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only