o-Xylene, Spectrophotometric Grade, 96% min
o-Xylene, Spectrophotometric Grade, 96% min
o-Xylene, Spectrophotometric Grade, 96% min
Thermo Scientific Chemicals

o-Xylene, Spectrophotometric Grade, 96% min

CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 032471.K2
also known as 032471-K2
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 30-Jun-2026
122.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS95-47-6
IUPAC Name1,2-xylene
Molecular FormulaC8H10
InChI KeyCTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on Evaporation5ppm
Residual water0.05%
o-Xylene is used as a solvent for paints and adhesives, and to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only