Dicyclopentadiene, typically 95%, stab.
Dicyclopentadiene, typically 95%, stab.
Dicyclopentadiene, typically 95%, stab.
Thermo Scientific Chemicals

Dicyclopentadiene, typically 95%, stab.

CAS: 77-73-6 | C10H12 | 132.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 032546.A1
also known as 032546-A1
Price (USD)
62.65
Online Exclusive
69.30
Save 6.65 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
69.30
Save 6.65 (10%)
Each
Chemical Identifiers
CAS77-73-6
IUPAC Nametricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular FormulaC10H12
InChI KeyHECLRDQVFMWTQS-UHFFFAOYNA-N
SMILESC1C=CC2C3CC(C=C3)C12
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤20
Impurity contentAcyclic Dienes: ≤2.0%
Impurity contentBHT: ≤0.04%
Impurity contentC9, C10, C11 codimers: ≤2.0%
Water Content (Karl Fischer Titration)≤300ppm
View more
Dicyclopentadiene is used in resins, particularly unsaturated polyester resins. It plays a major role in inks, adhesives and paints. It is also used as a monomer in polymerization reactions. It is a precursor for the preparation of endo-tetrahydrodicyclopentadiene, which reacts with aluminum chloride at higher temperature to give adamantine. It undergoes a retro-Diels-Alder reaction to yield cyclopentadiene, which acts as ligand in inorganic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only