Potassium borohydride, 98%
Potassium borohydride, 98%
Potassium borohydride, 98%
Thermo Scientific Chemicals

Potassium borohydride, 98%

Source of Hydrogen, Reducing Agent For Aldehydes, Ketones | CAS: 13762-51-1 | BH4K | 53.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
1 kg
Catalog number 032570.36
also known as 032570-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS13762-51-1
IUPAC Namepotassium boranuide
Molecular FormulaBH4K
InChI KeyICRGAIPBTSPUEX-UHFFFAOYSA-N
SMILES[BH4-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min
Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plasticsPotassium borohydride is used as a selective reducing agent for ketones, aldehydes and acid chlorides. It is used as a precursor in the production of chloramphenicol, vitamin A, thiopenicol, atropine and scopolamine. It is used to prepare high dispersive copper nanoparticles by the chemical reduction method using potassium borohydride as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only