Bromophenol Blue, ACS
Bromophenol Blue, ACS
Bromophenol Blue, ACS
Thermo Scientific Chemicals

Bromophenol Blue, ACS

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 032641.06
also known as 032641-06
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.70
Save 6.05 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.70
Save 6.05 (15%)
Each
Chemical Identifiers
CAS115-39-9
IUPAC Name3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H10Br4O5S
InChI KeyUDSAIICHUKSCKT-UHFFFAOYSA-N
SMILESOC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentVisual transition interval pH 3.0 (yellow) to pH 4.6 (blue)
CommentClarity of solution P.T.
Bromophenol blue is used as a tracking dye in electrophoresis, as a color marker in gel electrophoresis and polyacrylamide gel electrophoresis. As a biological stain, it plays an important role to stain proteins and nucleic acids. It acts an intermediate and an acid-base indicator in the pH range 3.0 to 4.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only