Cyanogen bromide
Cyanogen bromide
Cyanogen bromide
Thermo Scientific Chemicals

Cyanogen bromide

CAS: 506-68-3 | CBrN | 105.922 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 032731.06
also known as 032731-06
Price (USD)
39.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.80
Each
Chemical Identifiers
CAS506-68-3
IUPAC Namecarbononitridic bromide
Molecular FormulaCBrN
InChI KeyATDGTVJJHBUTRL-UHFFFAOYSA-N
SMILESBrC#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline granules or powder
Assay from Supplier's CofA≥96.0 to ≤104.0% (Titration)
Appearance (Color)Colorless to white
Cyanogen bromide is used as a reagent in molecular biology, peptide mapping, gold extraction and amino acid sequence analysis. It acts as a coupling reagent for phosphoramidate or pyrophosphate inter-nucleotide bonds in DNA duplexes. It is also used in organic synthesis to prepare cyanamides, dicyanamides, cyanuric bromide, guanidines and hydroxylguanidines. It is involved in the von Braun reaction, it gives disubstituted cyanamides by reacting with tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only