alpha,alpha-Dimethylphenylacetic acid
alpha,alpha-Dimethylphenylacetic acid
alpha,alpha-Dimethylphenylacetic acid
Thermo Scientific Chemicals

alpha,alpha-Dimethylphenylacetic acid

CAS: 826-55-1 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 032884.04
also known as 032884-04
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS826-55-1
IUPAC Name2-methyl-2-phenylpropanoic acid
Molecular FormulaC10H12O2
InChI KeyYYEROYLAYAVZNW-UHFFFAOYSA-N
SMILESCC(C)(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
Assay from Supplier's CofA≥97.0%
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
α,α-Dimethylphenylacetic acid is used in the synthesis of active pharmaceutical ingredient, fexofenadine hydrochloride, which is used as antihistamine. Further, it acts as pharmaceutical intermediates and as chemical intermediates in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only